Få mere at vide
1-NM-PP1, MedChemExpress
Artikelnummer. 16492067
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16492067 50 mg -
16478557 5 mg 5mg
16405928 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16492067 Leverandør Medchem Express Leverandørnr. HY13942
MedChemExpress 1-NM-PP1, a cell-permeable PP1 analog, is a potent Src family kinases inhibitor with IC50s of 4.3 nM and 3.2 nM for v-Src-as1 and c-Fyn-as1, respectively.
1-NM-PP1, a cell-permeable PP1 analog, is a potent Src family kinases inhibitor with IC50s of 4.3 nM and 3.2 nM for v-Src-as1 and c-Fyn-as1, respectively.
Tekniske data
Kemisk navn eller materiale 1-NM-PP1
Fysisk form Solid
CAS 221244-14-0
Farve Off-White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C20H21N5
Til brug med (applikation) Cancer-Kinase/protease
Synonym PP1 Analog II
Opløselighedsinformation DMSO : 27.5 mg/mL (82.98 mM; Need ultrasonic and warming)
SMIL CC(N1N=C(CC2=CC=CC3=C2C=CC=C3)C4=C(N)N=CN=C41)(C)C
Molekylvægt (g/mol) 331.41
Formel vægt 331.41
Procent renhed 99.28%