missing translation for 'onlineSavingsMsg'
Learn More

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog Number p-7080885
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
131-91-9
C10H7NO2
173.171
MFCD00003884
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
8580
1-nitrosonaphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2N=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
131-91-9
C10H7NO2
173.171
MFCD00003884
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
8580
1-nitrosonaphthalen-2-ol
C1=CC=C2C(=C1)C=CC(=C2N=O)O

1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Nitroso-2-naphthol as a complexing agent for the on-line preconcentration of cobalt with FI-FAAS. 1-Nitroso-2-naphthol is the chelating ion-exchanger in the synthesis of alumina adsorbents that are of acidic, basic and neutral nature. It is used for the removal and preconcentration of Pb(II), Cu(II), Cr(III) from waste, as well as drinking water. It is also used in the preconcentration of cobalt using C(18) microcolumn with flow injection that is coupled to a flame atomic absorption spectrometry (FI-FAAS) system. 1-Nitroso-2-naphthol is used in the preparation of 1-nitroso-2-naphthol-sodium nitrite reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 104°C to 108°C (decomposition)
Quantity 10 g
UN Number UN3077
Beilstein 776947
Merck Index 14,6641
Solubility Information Insoluble in water.
Formula Weight 173.17
Percent Purity 99%
Chemical Name or Material 1-Nitroso-2-naphthol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'