Få mere at vide
1-Nitropyrene-d9, TRC
Artikelnummer. 30464951
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30464951 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464951 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5199522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Nitropyrene-d9
CAS 93487-20-8
Molekylær formel C16 2H9 N O2
Mængde 2.5 mg
Synonym Pyrene-1,2,3,4,5,6,7,9,10-d9, 8-nitro-, 8-Nitropyrene-1,2,3,4,5,6,7,9,10-d9, 1-Nitropyrene-d9, 1-Nitropyrene D9
InChI formel InChI=1S/C16H9NO2/c18-17(19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMIL [2H]c1c([2H])c2c([2H])c([2H])c3c([2H])c([2H])c(c4c([2H])c([2H])c(c1[2H])c2c34)[N+](=O)[O-]
IUPAC navn 1,2,3,4,5,6,7,9,10-nonadeuterio-8-nitropyrene
Molekylvægt (g/mol) 256.3
Formel vægt 256.1198