Få mere at vide
1-Nitropyrene-d9, CDN
Artikelnummer. 30321511
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321511 0.05 g
30319289 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321511 Leverandør CDN Leverandørnr. CDND52340.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Nitropyrene-d9
CAS 93487-20-8
Molekylær formel C16 2H9 N O2
Mængde 0.05 g
Synonym Pyrene-1,2,3,4,5,6,7,9,10-d9, 8-nitro-, 8-Nitropyrene-1,2,3,4,5,6,7,9,10-d9, 1-Nitropyrene-d9, 1-Nitropyrene D9
InChI formel InChI=1 S/C16H9NO2/c18-17(19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D
SMIL [2 H]c1c([2 H])c2c([2 H])c([2 H])c3c([2 H])c([2 H])c(c4c([2 H])c([2 H])c(c1[2 H])c2c34)[N+](=O)[O-]
IUPAC navn 1,2,3,4,5,6,7,9,10-nonadeuterio-8-nitropyrene
Molekylvægt (g/mol) 256.3
Formel vægt 256.1198 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity