Få mere at vide
1-Nitronaphthalene-d7, TRC
Artikelnummer. 30481782
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481782 10 mg
30475474 1 mg
30471819 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481782 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49685610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 80789-77-1
Molekylær formel C10 D7 N O2
Molekylvægt (g/mol) 180.211
Synonym Naphthalene-1,2,3,4,5,6,7-d7, 8-nitro- (9CI), 1-Nitronaphthalene-d7, 1-Nitronaphthalene-2,3,4,5,6,7,8-d7, 8-Nitronaphthalene-1,2,3,4,5,6,7-d7, 1-Nitronaphthalene-D7
IUPAC navn 1,2,3,4,5,6,7-heptadeuterio-8-nitronaphthalene
SMIL [2H]c1c([2H])c([2H])c2c(c([2H])c([2H])c([2H])c2c1[2H])[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 1-Nitronaphthalene-d7
Mængde 10 mg
InChI formel InChI=1S/C10H7NO2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H/i1D,2D,3D,4D,5D,6D,7D
Formel vægt 180.092