missing translation for 'onlineSavingsMsg'
Learn More
1-Nitro-3,5-bis(trifluoromethyl)benzene, 96% Product Code.: 11422828

1-Nitro-3,5-bis(trifluoromethyl)benzene, 96%

Product Code. p-7023350
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11422828

Brand: Thermo Scientific Alfa Aesar A16797.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 328-75-6 | C8H3F6NO2 | 259.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-75-6
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.11
MDL Number MFCD00000384
InChI Key GMUWJDVVXLBMEZ-UHFFFAOYSA-N
Synonym 1-nitro-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl nitrobenzene, 1,3-bis trifluoromethyl-5-nitrobenzene, 3,5-di trifluoromethyl nitrobenzene, benzene, 1-nitro-3,5-bis trifluoromethyl, 3,5-bis-trifluoromethylnitrobenzene, 3,5-di trifluoromethylnitro benzene, 3,5-bis-trifluoromethyl nitrobenzene, 5-nitro-1,3-bis trifluoromethyl benzene, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-5-nitroxylene
PubChem CID 67603
IUPAC Name 1-nitro-3,5-bis(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.535
Boiling Point 75°C to 76°C (11 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.427
Quantity 5 g
Beilstein 1998251
Formula Weight 259.11
Percent Purity ≥97%
Chemical Name or Material 1-Nitro-3,5-bis(trifluoromethyl)benzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10254362 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeA16797.14,A16797.06
Lot Number10248236 Certificate TypeCertificate of Analysis Date06/10/2023 Product CodeA16797.14,A16797.06
Lot Number10245400 Certificate TypeCertificate of Analysis Date17/01/2023 Product CodeA16797.14,A16797.06
Lot Number10148146 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA16797.14,A16797.06
Lot Number10180527 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA16797.14,A16797.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Nitro-3,5-bis(trifluoromethyl)benzene, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.