Få mere at vide
1-Naphthylhydrazine, TRC
Artikelnummer. 30475920
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475920 10 mg
30474433 100 mg
30472667 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475920 Leverandør Toronto Research Chemicals Leverandørnr. TRCN36852810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2243-55-2
Molekylær formel C10H10N2
Molekylvægt (g/mol) 158.2
IUPAC navn naphthalen-1-ylhydrazine
SMIL NNc1cccc2ccccc12
Tekniske data
Kemisk navn eller materiale 1-Naphthylhydrazine
Mængde 10 mg
InChI formel InChI=1S/C10H10N2/c11-12-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,12H,11H2
Formel vægt 158.0844