Få mere at vide
1-Naphthyl PP1 hydrochloride, MedChemExpress
Artikelnummer. 16495918
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16495918 10 mg 10mg
16482067 50 mg -
16468557 5 mg -
16436366 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16495918 Leverandør Medchem Express Leverandørnr. HY13941B
MedChemExpress 1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.
1-Naphthyl PP1 hydrochloride (1-NA-PP 1 hydrochloride) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 hydrochloride inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.
Tekniske data
Kemisk navn eller materiale 1-Naphthyl PP1 hydrochloride
Fysisk form Solid
CAS 956025-47-1
Farve White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C19H20ClN5
Til brug med (applikation) Cancer-Kinase/protease
Synonym 1-NA-PP 1 hydrochloride
Opløselighedsinformation DMSO : ≥ 73 mg/mL (206.30 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL CC(N1N=C(C2=C3C=CC=CC3=CC=C2)C4=C(N)N=CN=C41)(C)C.[H]Cl
Molekylvægt (g/mol) 353.85
Formel vægt 353.85
Procent renhed 99.94%