Learn More
1-Naphthol, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | 1-Naphthol |
| CAS | 90-15-3 |
| Density | 1.28 g/cm³ at 20°C |
| Boiling Point | 288°C (1013 hPa) |
| Flash Point | 125°C (closed cup) |
| Melting Range | 94°C to 96°C |
| Quantity | 50 kg |
| Synonym | 1-Naphthol, 1-Hydroxynaphthalene, 1-Naphthol |
| Molecular Weight (g/mol) | 144.17 |
| Grade | Not Available |