Få mere at vide
1-Naphthoic Acid-d7, TRC
Artikelnummer. 30476039
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30476039 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476039 Leverandør Toronto Research Chemicals Leverandørnr. TRCN3452521G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Naphthoic Acid-d7
CAS 634179-80-9
Molekylær formel C11 2H7 H O2
Mængde 1 g
InChI formel InChI=1S/C11H8O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H,12,13)/i1D,2D,3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c2c(C(=O)O)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn 2,3,4,5,6,7,8-heptadeuterionaphthalene-1-carboxylic acid
Molekylvægt (g/mol) 179.22
Formel vægt 179.0964