Få mere at vide
1-Naphthalenemethanol-d7, TRC
Artikelnummer. 30479752
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30479752 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479752 Leverandør Toronto Research Chemicals Leverandørnr. TRCN372552500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Naphthalenemethanol-d7
CAS 1189876-86-5
Molekylær formel C11H3D7O
Mængde 500 mg
Synonym α-Naphthylmethanol-d7, α-Naphthylcarbinol-d7, Naphthalen-1-ylmethanol-d7, NSC 5315-d7
InChI formel InChI=1S/C11H10O/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7,12H,8H2/i1D,2D,3D,4D,5D,6D,7D
SMIL [2H]C1=C([2H])C([2H])=C2C(C([2H])=C([2H])C([2H])=C2[2H])=C1CO
IUPAC navn (naphthalen-1-yl-d7)methanol
Molekylvægt (g/mol) 165.24
Formel vægt 165.12