missing translation for 'onlineSavingsMsg'
Learn More

1-n-Octylpiperazine, 97%, Thermo Scientific™

Catalog Number 15471988
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
54256-45-0
C12H26N2
198.354
MFCD00958407
RRZDZQLIIMBIDL-UHFFFAOYSA-N
1-n-octylpiperazine, 1-1-octyl-piperazine, 1-n-octyl-piperazine, 1-octyl-piperazine, 1-n-octyl piperazine, piperazine, 1-octyl, 1-1-octyl piperazine, 1-oct-1-yl piperazin, 1-octylpiperazine
2737099
1-octylpiperazine
CCCCCCCCN1CCNCC1
This item is not returnable. View return policy
54256-45-0
C12H26N2
198.354
MFCD00958407
RRZDZQLIIMBIDL-UHFFFAOYSA-N
1-n-octylpiperazine, 1-1-octyl-piperazine, 1-n-octyl-piperazine, 1-octyl-piperazine, 1-n-octyl piperazine, piperazine, 1-octyl, 1-1-octyl piperazine, 1-oct-1-yl piperazin, 1-octylpiperazine
2737099
1-octylpiperazine
CCCCCCCCN1CCNCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.836 g/mL
Boiling Point 240°C to 241°C
Flash Point 46°C (115°F)
Refractive Index 1.467
Quantity 250 mg
UN Number UN1993
Formula Weight 198.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-n-Octylpiperazine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'