Få mere at vide
1-Methyluric Acid-d3, TRC
Artikelnummer. 30483385
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483385 10 mg
30467655 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483385 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33818210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189480-64-5
Molekylær formel C6 2H3 H3 N4 O3
Molekylvægt (g/mol) 185.16
IUPAC navn 1-(trideuteriomethyl)-7,9-dihydro-3H-purine-2,6,8-trione
SMIL [2H]C([2H])([2H])N1C(=O)NC2=C(NC(=O)N2)C1=O
Tekniske data
Kemisk navn eller materiale 1-Methyluric Acid-d3
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C6H6N4O3/c1-10-4(11)2-3(9-6(10)13)8-5(12)7-2/h1H3,(H,9,13)(H2,7,8,12)/i1D3
Formel vægt 185.0628
Procent renhed >95%