Få mere at vide
1-Methylpyrrolidine-d8, TRC
Artikelnummer. 30483860
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30483860 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483860 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32621150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methylpyrrolidine-d8
CAS 131857-29-9
Molekylær formel C5H3D8N
Mængde 50 mg
Synonym N-Methylpyrrolidine-d8, N-Methyltetrahydropyrrole-d8, NSC 65579-d8, N-Methylpyrrolidine-2,2,3,3,4,4,5,5-d8
InChI formel InChI=1S/C5H11N/c1-6-4-2-3-5-6/h2-5H2,1H3/i2D2,3D2,4D2,5D2
SMIL [2H]C1([2H])N(C)C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 2,2,3,3,4,4,5,5-octadeuterio-1-methylpyrrolidine
Molekylvægt (g/mol) 93.2
Formel vægt 93.1394