Få mere at vide
1-Methylpyrrolidin-3-yl 2-Cyclopentyl-2-hydroxy-2-phenylacetate, Mikromol™
Beskrivelse
1-Methylpyrrolidin-3-yl 2-Cyclopentyl-2-hydroxy-2-phenylacetate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-Methylpyrrolidin-3-yl 2-Cyclopentyl-2-hydroxy-2-phenylacetate |
| CAS | 13118-11-1 |
| Analyt- eller komponentnavne | 1-Methylpyrrolidin-3-yl 2-Cyclopentyl-2-hydroxy-2-phenylacetate |
| Forsendelsestilstand | Ice pack (-20°C) |
| Molekylær formel | C18 H25 N O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Glycopyrronium Bromide Imp. G (EP), Glycopyrrolate USP Related Compound B, Glycopyrrolate USP RC B, Glycopyrrolate Related Compound B as free base, Glycopyrrolate Related Compound B, Glycopyrronium Bromide Impurity G, Glycopyrronium Impurity G |
| InChI formel | InChI=1S/C18H25NO3/c1-19-12-11-16(13-19)22-17(20)18(21,15-9-5-6-10-15)14-7-3-2-4-8-14/h2-4,7-8,15-16,21H,5-6,9-13H2,1H3 |
| SMIL | CN1CCC(C1)OC(=O)C(O)(C2CCCC2)c3ccccc3 |
| IUPAC navn | (1-methylpyrrolidin-3-yl) 2-cyclopentyl-2-hydroxy-2-phenylacetate |
| Vis mere |