Få mere at vide
1-Methylpropyl Ester Methanesulfonic Acid, TRC
Artikelnummer. 30337979
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30337979 50 mg
30336781 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30337979 Leverandør Toronto Research Chemicals Leverandørnr. TRCB48561050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methylpropyl Ester Methanesulfonic Acid
CAS 16156-54-0
Molekylær formel C5H12O3S
Mængde 50 mg
InChI formel InChI=1S/C5H12O3S/c1-4-5(2)8-9(3,6)7/h5H,4H2,1-3H3
SMIL CCC(C)OS(=O)(=O)C
IUPAC navn butan-2-yl methanesulfonate
Molekylvægt (g/mol) 152.21
Formel vægt 152.0507