Få mere at vide
1-Methylpiperidin-3-one, TRC
Artikelnummer. 30478522
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30478522 100 mg
30472586 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30478522 Leverandør Toronto Research Chemicals Leverandørnr. TRCM320983100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5519-50-6
Molekylær formel C6H11NO
Molekylvægt (g/mol) 113.15
IUPAC navn 1-methylpiperidin-3-one
SMIL CN1CCCC(=O)C1
Tekniske data
Kemisk navn eller materiale 1-Methylpiperidin-3-one
Mængde 100 mg
InChI formel InChI=1S/C6H11NO/c1-7-4-2-3-6(8)5-7/h2-5H2,1H3
Formel vægt 113.0841