Få mere at vide
1-Methylpiperazine-2,2,3,3,5,5,6,6-d8, TRC
Artikelnummer. 30473999
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30473999 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473999 Leverandør Toronto Research Chemicals Leverandørnr. TRCM91735025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methylpiperazine-2,2,3,3,5,5,6,6-d8
CAS 917358-65-7
Molekylær formel C5 2H8 H4 N2
Mængde 25 mg
Synonym 1-Methyl-1,4-piperazine-d8, N-Methylpiperazine-d8, NSC 30195-d8, NSC 30675-d8, Imatinib EP Impurity G-d8
InChI formel InChI=1S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3/i2D2,3D2,4D2,5D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(C)C1([2H])[2H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuterio-1-methylpiperazine
Molekylvægt (g/mol) 108.212
Formel vægt 108.15