Få mere at vide
1-Methylnicotinamide Iodide, LoGiCal
Artikelnummer. 30461399
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461399 10 mg
1 options
Artikelnummer. 30461399 Leverandør LoGiCal Leverandørnr. LGCFOR0552.05
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methylnicotinamide Iodide
CAS 6456-44-6
Analyt- eller komponentnavne 1-Methylnicotinamide Iodide
Forsendelsestilstand Room Temperature
Molekylær formel C7 H9 N2 O . I
Certificeringer/compliance ISO 9001
Synonym Pyridinium, 3-(aminocarbonyl)-1-methyl-, iodide (1:1), 3-Carbamoyl-1-methylpyridinium iodide (6CI,7CI), Pyridinium, 3-(aminocarbonyl)-1-methyl-, iodide (9CI), Pyridinium, 3-carbamoyl-1-methyl-, iodide (8CI), Pyridinium, 3-carbamyl-1-methyl-, iodide (4CI), 1-Methylnicotinamide iodide, Nicotinamide methiodide
InChI formel InChI=1S/C7H8N2O.HI/c1-9-4-2-3-6(5-9)7(8)10;/h2-5H,1H3,(H-,8,10);1H
Løsningstype Neat
SMIL [I-].C[n+]1cccc(c1)C(=O)N
IUPAC navn 1-methylpyridin-1-ium-3-carboxamide;iodide
Industry Type Clinical Testing
Molekylvægt (g/mol) 264.06
Mængde 10 mg
Formel vægt 263.976 g/mol