Få mere at vide
1-Methylinosine, MedChemExpress
Artikelnummer. 16431037
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16431037 50 mg
16424758 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16431037 Leverandør Medchem Express Leverandørnr. HY11313950mg
MedChemExpress 1-Methylinosine is a modified nucleotide found at position 37 in tRNA 3' to the anticodon of eukaryotic tRNA.
1-Methylinosine is a modified nucleotide found at position 37 in tRNA 3' to the anticodon of eukaryotic tRNA.
Tekniske data
Kemisk navn eller materiale 1-Methylinosine
CAS 2140-73-0
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C11H14N4O5
Mængde 50 mg
Synonym N1-Methylinosine
Opløselighedsinformation DMSO : 20.83 mg/mL (73.80 mM; Need ultrasonic)
SMIL CN1C=NC(N([C@H]2[C@H](O)[C@H](O)[C@@H](CO)O2)C=N3)=C3C1=O
Molekylvægt (g/mol) 282.25
Formel vægt 282.25
Procent renhed 99.89%
Grad Research
Fysisk form Powder
Til brug med (applikation) Metabolism-protein/nucleotide metabolism