missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindoline-5-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-3586614
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
60082-02-2
C10H11NO
161.204
MFCD03208513
OZQGVTARHLDSTI-UHFFFAOYSA-N
1-methylindoline-5-carbaldehyde, 1-methyl-2,3-dihydro-1h-indole-5-carbaldehyde, 1-methylindoline-5-carboxaldehyde, 1-methyl-5-indolinecarbaldehyde, 1-methyl-5-formylindoline, 5-formyl-1-methylindoline, 1-methyl-5-indolinecarbaldehyde #, 2,3-dihydro-1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde, 2,3-dihydro-1-methyl
595181
1-methyl-2,3-dihydroindole-5-carbaldehyde
CN1CCC2=C1C=CC(=C2)C=O
This item is not returnable. View return policy
60082-02-2
C10H11NO
161.204
MFCD03208513
OZQGVTARHLDSTI-UHFFFAOYSA-N
1-methylindoline-5-carbaldehyde, 1-methyl-2,3-dihydro-1h-indole-5-carbaldehyde, 1-methylindoline-5-carboxaldehyde, 1-methyl-5-indolinecarbaldehyde, 1-methyl-5-formylindoline, 5-formyl-1-methylindoline, 1-methyl-5-indolinecarbaldehyde #, 2,3-dihydro-1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde, 2,3-dihydro-1-methyl
595181
1-methyl-2,3-dihydroindole-5-carbaldehyde
CN1CCC2=C1C=CC(=C2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material 1-Methylindoline-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'