Learn More
1-Methylindole-2-carboxylic acid, Thermo Scientific Chemicals
Catalog Number p-4089828
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
16136-58-6
C10H9NO2
175.19
MFCD00005801
MAHAMBLNIDMREX-UHFFFAOYSA-N
1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa
27695
1-methylindole-2-carboxylic acid
CN1C2=CC=CC=C2C=C1C(=O)O
16136-58-6
C10H9NO2
175.19
MFCD00005801
MAHAMBLNIDMREX-UHFFFAOYSA-N
1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa
27695
1-methylindole-2-carboxylic acid
CN1C2=CC=CC=C2C=C1C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 212°C to 213°C
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Beilstein 22, V,3, 15
Formula Weight 175.19
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylindole-2-carboxylic acid
compliance-icons
Product Identifier
  • 1-Methylindole-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.