Få mere at vide
1-Methylimidazole-d3 (ring-d3), TRC
Artikelnummer. 30476729
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476729 50 mg
30477057 5 mg
30471760 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476729 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31298150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4166-68-1
Molekylær formel C4 D3 H3 N2
Molekylvægt (g/mol) 85.122
Synonym 1H-Imidazole-2,4,5-d3, 1-methyl- (9CI, ACI), 1-Methyl-1H-imidazole-2,4,5-d3 (ACI), Imidazole-2,4,5-d3, 1-methyl- (7CI, 8CI), 1-Methylimidazole-2,4,5-d3, N-Methylimidazole-d3, 1-Methyl-1H-imidazole-2,4,5-D3, 1-Methylimidazole-d3, 1-Methylimidazole-d3 (ring-d3)
IUPAC navn 2,4,5-trideuterio-1-methylimidazole
SMIL [2H]c1nc([2H])n(C)c1[2H]
Tekniske data
Kemisk navn eller materiale 1-Methylimidazole-d3 (ring-d3)
Mængde 50 mg
InChI formel InChI=1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3/i2D,3D,4D
Formel vægt 85.072