Learn More
1-Methylimidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Air sensitive. Store in a cool, dry place. Store in a tightly closed container. Store under nitrogen.
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Air sensitive. Store in a cool, dry place. Store in a tightly closed container. Store under nitrogen.
Specifications
Melting Point | 36°C to 39°C |
Boiling Point | 70°C to 74°C (1 mmHg) |
Flash Point | 102°C (216°F) |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 110.12 |
Percent Purity | 98% |
Chemical Name or Material | 1-Methylimidazole-2-carboxaldehyde |