Få mere at vide
1-Methylhydantoin, TRC
Artikelnummer. 30483629
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30483629 2.5 g 2.5g
30469304 250 mg -
30483392 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483629 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2196802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 616-04-6
Molekylær formel C4H6N2O2
Molekylvægt (g/mol) 114.1
Synonym Hydantoin, 1-methyl- (6CI,7CI,8CI), 1-Methyl-2,4-imidazolidinedione, 1-Methylimidazolidine-2,4-dione, Dioxy-creatinine, NSC 80560
IUPAC navn 1-methylimidazolidine-2,4-dione
SMIL CN1CC(=O)NC1=O
Tekniske data
Kemisk navn eller materiale 1-Methylhydantoin
Mængde 2.5 g
InChI formel InChI=1S/C4H6N2O2/c1-6-2-3(7)5-4(6)8/h2H2,1H3,(H,5,7,8)
Formel vægt 114.0429