missing translation for 'onlineSavingsMsg'
Få mere at vide

(1-Methylethyl)carbamic Acid-d7 Phenyl Ester, TRC

Artikelnummer. 30479435
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479435 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479435 Leverandør Toronto Research Chemicals Leverandørnr. TRCM304912100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (1-Methylethyl)carbamic Acid-d7 Phenyl Ester
CAS 1329835-22-4
Molekylær formel C10 D7 H6 N O2
Mængde 100 mg
Synonym Phenyl N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)carbamate, Isopropyl-d7-carbamic acid phenyl ester, Phenyl N-[1-(methyl-d3)ethyl-1,2,2,2-d4]carbamate, Phenyl N-(isopropyl-d7)carbamate, [1-(Methyl-d3)ethyl-1,2,2,2-d4]carbamic acid phenyl ester, Phenyl isopropylcarbamate-D7 (isopropyl-D7)
InChI formel InChI=1S/C10H13NO2/c1-8(2)11-10(12)13-9-6-4-3-5-7-9/h3-8H,1-2H3,(H,11,12)/i1D3,2D3,8D
SMIL [2H]C([2H])([2H])C([2H])(NC(=O)Oc1ccccc1)C([2H])([2H])[2H]
IUPAC navn phenyl N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)carbamate
Molekylvægt (g/mol) 186.259
Formel vægt 186.139
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.