Få mere at vide
1-[(Methylamino)[[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]amino]methylene]urea Dihydrochloride (Cimetidine Amide Dihydrochloride), Mikromol™
Beskrivelse
1-[(Methylamino)[[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]amino]methylene]urea Dihydrochloride (Cimetidine Amide Dihydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cimetidine Amide Dihydrochloride |
| CAS | 52568-80-6 |
| Analyt- eller komponentnavne | Cimetidine Amide Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H18 N6 O S . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Cimetidine Hydrochloride Imp. C (EP), Cimetidine Imp. C (EP), 1-[(Methylamino)[[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]amino]methylene]urea Dihydrochloride, Cimetidine Hydrochloride Imp. C (EP) as Dihydrochloride, Cimetidine Amide Dihydrochloride, Cimetidine Imp. C (EP) as Dihydrochloride, Ph Eur Cimetidine Impurity C, SKF-92422-A2, Cimetidine Hydrochloride Impurity C, Cimetidine Impurity C as Dihydrochloride, Cimetidine Impurity C |
| InChI formel | InChI=1S/C10H18N6OS.2ClH/c1-7-8(15-6-14-7)5-18-4-3-13-10(12-2)16-9(11)17;;/h6H,3-5H2,1-2H3,(H,14,15)(H4,11,12,13,16,17);2*1H |
| SMIL | Cl.Cl.CNC(=NC(=O)N)NCCSCc1nc[nH]c1C |
| IUPAC navn | [methylamino-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethylamino]methylidene]urea;dihydrochloride |
| Vis mere |