Få mere at vide
1-Methyl-nicotinamide Iodide, TRC
Artikelnummer. 30240619
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240619 250 mg
30240914 500 mg
30240896 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240619 Leverandør Toronto Research Chemicals Leverandørnr. TRCM323235250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6456-44-6
Molekylær formel C7H9IN2O
Molekylvægt (g/mol) 264.06
Synonym Pyridinium, 3-(aminocarbonyl)-1-methyl-, iodide (1:1), 3-Carbamoyl-1-methylpyridinium iodide (6CI,7CI), Pyridinium, 3-(aminocarbonyl)-1-methyl-, iodide (9CI), Pyridinium, 3-carbamoyl-1-methyl-, iodide (8CI), Pyridinium, 3-carbamyl-1-methyl-, iodide (4CI), 1-Methylnicotinamide iodide, Nicotinamide methiodide
IUPAC navn 1-methylpyridin-1-ium-3-carboxamide;iodide
SMIL [I-].C[n+]1cccc(c1)C(=O)N
Tekniske data
Kemisk navn eller materiale 1-Methyl-nicotinamide Iodide
Mængde 250 mg
InChI formel InChI=1S/C7H8N2O.HI/c1-9-4-2-3-6(5-9)7(8)10;/h2-5H,1H3,(H-,8,10);1H
Formel vægt 263.976