missing translation for 'onlineSavingsMsg'
Få mere at vide

1-Methyl-L-histidine, TRC

Artikelnummer. 30132483
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132483 250 mg
30010321 10 mg
30008626 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30132483 Leverandør Toronto Research Chemicals Leverandørnr. TRCM312050250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 332-80-9
Molekylær formel C7 H11 N3 O2
Molekylvægt (g/mol) 169.18
Synonym 1-Methyl-L-histidine, Histidine, 1-methyl-, L- (8CI), 1-Methyl-L-histidine, (S)-2-Amino-3-(1-methyl-1H-imidazol-4-yl)propanoic acid, 1-Methylhistidine, 1-N-Methyl-L-histidine, N1-Methyl-L-histidine, NSC 524367
IUPAC navn (2S)-2-amino-3-(1-methylimidazol-4-yl)propanoic acid
SMIL Cn1cnc(C[C@H](N)C(=O)O)c1

Specifications

Kemisk navn eller materiale 1-Methyl-L-Histidine
InChI formel InChI=1S/C7H11N3O2/c1-10-3-5(9-4-10)2-6(8)7(11)12/h3-4,6H,2,8H2,1H3,(H,11,12)/t6-/m0/s1
Mængde 250 mg
Formel vægt 169.0851
Forsendelsestilstand Dry Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.