Få mere at vide
1-Methyl-ethyl 4-Methyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxylate Hydrochloride (Articaine Isopropyl Ester Hydrochloride), Mikromol™
Beskrivelse
1-Methyl-ethyl 4-Methyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxylate Hydrochloride (Articaine Isopropyl Ester Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Articaine Isopropyl Ester Hydrochloride |
| CAS | 2984093-98-1 |
| Analyt- eller komponentnavne | Articaine Isopropyl Ester Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H24 N2 O3 S . Cl H |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | Articaine Hydrochloride Imp. C (EP), Articaine Imp. C (EP), 1-Methyl-ethyl 4-Methyl-3-[[(2RS)-2-(propylamino)propanoyl]amino]thiophene-2-carboxylate Hydrochloride, Articaine Hydrochloride Imp. C (EP) as Hydrochloride, Articaine Isopropyl Ester Hydrochloride, Articaine Hydrochloride Impurity C as Hydrochloride, Articaine Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C16H16O2/c1-12(18-2)14-9-6-10-15(11-14)16(17)13-7-4-3-5-8-13/h3-12H,1-2H4 |
| SMIL | Cl.CCCNC(C)C(=O)Nc1c(C)csc1C(=O)OC(C)C |
| IUPAC navn | propan-2-yl 4-methyl-3-[2-(propylamino)propanoylamino]thiophene-2-carboxylate;hydrochloride |
| Vis mere |