Få mere at vide
1-Methyl-d3-L-tryptophan, CDN
Artikelnummer. 30324768
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324768 0.05 g
30315047 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324768 Leverandør CDN Leverandørnr. CDND69710.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-L-tryptophan-D3 (1-methyl-D3)
CAS 1801851-87-5
Molekylær formel C12 D3 H11 N2 O2
Mængde 0.05 g
Synonym L-Tryptophan, 1-(methyl-d3)- (ACI), 1-(Methyl-d3)-L-tryptophan (ACI), 1-Methyl-L-tryptophan-d3, 1-Methyl-L-tryptophan-D3 (1-methyl-D3), (2 S)-2-Amino-3-[1-(methyl-d3)-1 H-indol-3-yl]propanoic acid, 1-Methyl-d3-L-tryptophan
InChI formel InChI=1 S/C12H14N2O2/c1-14-7-8(6-10(13)12(15)16)9-4-2-3-5-11(9)14/h2-5,7,10 H,6,13H2,1H3,(H,15,16)/t10-/m0/s1/i1D3
SMIL [2 H]C([2 H])([2 H])n1cc(C[C@H](N)C(=O)O)c2ccccc12
IUPAC navn (2 S)-2-amino-3-[1-(trideuteriomethyl)indol-3-yl]propanoic acid
Molekylvægt (g/mol) 221.2703
Formel vægt 221.1244 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity