Få mere at vide
1-Methyl-d3-imidazole, CDN
Artikelnummer. 30317104
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317104 0.25 g
30309997 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317104 Leverandør CDN Leverandørnr. CDND69250.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-d3-imidazole
CAS 16650-76-3
Molekylær formel C4 2H3 H3 N2
Mængde 0.25 g
InChI formel InChI=1 S/C4H6N2/c1-6-3-2-5-4-6/h2-4 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])n1ccnc1
IUPAC navn 1-(trideuteriomethyl)imidazole
Molekylvægt (g/mol) 85.12
Formel vægt 85.0719 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity