Få mere at vide
1-Methyl-d3-5-mercapto-1,2,3,4-tetrazole, CDN
Artikelnummer. 30321474
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321474 0.05 g
30311263 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321474 Leverandør CDN Leverandørnr. CDND27970.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-5-tetrazolethiol methyl-d3
CAS 345909-96-8
Molekylær formel C2 2H3 H N4 S
Mængde 0.05 g
InChI formel InChI=1 S/C2H4N4S/c1-6-2(7)3-4-5-6/h1H3,(H,3,5,7)/i1D3
SMIL [2 H]C([2 H])([2 H])n1nnnc1S
IUPAC navn 1-(trideuteriomethyl)tetrazole-5-thiol
Molekylvægt (g/mol) 119.16
Formel vægt 119.0345 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity