Få mere at vide
1-(Methyl-D3)-4-nitro-3-propyl-1H-pyrazole-5-carboxylic Acid, TRC
Artikelnummer. 30481185
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30481185 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481185 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32053225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(Methyl-D3)-4-nitro-3-propyl-1H-pyrazole-5-carboxylic Acid
CAS 526203-29-2
Molekylær formel C8H8D3N3O4
Mængde 25 mg
InChI formel InChI=1S/C8H11N3O4/c1-3-4-5-6(11(14)15)7(8(12)13)10(2)9-5/h3-4H2,1-2H3,(H,12,13)/i2D3
SMIL [2H]C([2H])([2H])n1nc(CCC)c(c1C(=O)O)[N+](=O)[O-]
IUPAC navn 4-nitro-5-propyl-2-(trideuteriomethyl)pyrazole-3-carboxylic acid
Molekylvægt (g/mol) 216.21
Formel vægt 216.0938