Få mere at vide
1-Methyl-d3-2-(4-methylbenzoyl)pyrrole, TRC
Artikelnummer. 30479252
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30479252 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479252 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3204921G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-d3-2-(4-methylbenzoyl)pyrrole
CAS 1185001-74-4
Molekylær formel C10 H8 O6 S2 . C5 H11 N O2
Mængde 1 g
InChI formel InChI=1S/C10H8O6S2.C5H11NO2/c11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16;1-4(3-6)2-5(7)8/h1-6H,(H,11,12,13)(H,14,15,16);4H,2-3,6H2,1H3,(H,7,8)
SMIL CC(CN)CC(=O)O.OS(=O)(=O)c1cccc2c(cccc12)S(=O)(=O)O
IUPAC navn 4-amino-3-methylbutanoic acid;naphthalene-1,5-disulfonic acid
Molekylvægt (g/mol) 405.443
Formel vægt 405.055