missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-phenyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2696709
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
105994-75-0
C11H10N2O2
202.213
MFCD04038957
MXGOKNNGBUOAGF-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-methyl-5-phenyl, acmc-1c8ot, 1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenyl, 1ff
2795474
1-methyl-5-phenylpyrazole-4-carboxylic acid
CN1C(=C(C=N1)C(=O)O)C2=CC=CC=C2
This item is not returnable. View return policy
105994-75-0
C11H10N2O2
202.213
MFCD04038957
MXGOKNNGBUOAGF-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-methyl-5-phenyl, acmc-1c8ot, 1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenyl, 1ff
2795474
1-methyl-5-phenylpyrazole-4-carboxylic acid
CN1C(=C(C=N1)C(=O)O)C2=CC=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'