Få mere at vide
1-Methyl-5-nitro-1H-benzimidazole-2-butanoic Acid Ethyl Ester-d3, TRC
Artikelnummer. 30480189
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30480189 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480189 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32364725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-5-nitro-1H-benzimidazole-2-butanoic Acid Ethyl Ester-d3
CAS 1286893-94-4
Molekylær formel C14N14D3N3O4
Mængde 25 mg
Synonym 1-Methyl-5-nitro-2-benzimidazolebutyric Acid Ethyl Ester-d3
InChI formel InChI=1S/C14H17N3O4/c1-3-21-14(18)6-4-5-13-15-11-9-10(17(19)20)7-8-12(11)16(13)2/h7-9H,3-6H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])n1c(CCCC(=O)OCC)nc2cc(ccc12)[N+](=O)[O-]
IUPAC navn ethyl 4-[5-nitro-1-(trideuteriomethyl)benzimidazol-2-yl]butanoate
Molekylvægt (g/mol) 294.32
Formel vægt 294.1407