Få mere at vide
1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester-d3, TRC
Artikelnummer. 30483033
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30483033 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483033 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28564710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester-d3
CAS 1246819-70-4
Molekylær formel C14H16D3N3O2
Mængde 10 mg
Synonym 5-Amino-1-methyl-2-benzimidazolebutyric Acid Ethyl Ester-d3
InChI formel InChI=1S/C14H19N3O2/c1-3-19-14(18)6-4-5-13-16-11-9-10(15)7-8-12(11)17(13)2/h7-9H,3-6,15H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])n1c(CCCC(=O)OCC)nc2cc(N)ccc12
IUPAC navn ethyl 4-[5-amino-1-(trideuteriomethyl)benzimidazol-2-yl]butanoate
Molekylvægt (g/mol) 264.34
Formel vægt 264.1666