Learn More
1-Methyl-4-piperidone, 98%
Description
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.
Solubility
It is miscible with water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong acids.

Specifications
Specifications
Density | 0.973 |
pH | 12 |
Boiling Point | 55°C to 58°C (10 mmHg) |
Flash Point | 60°C (140°F) |
Odor | Amine-like |
Refractive Index | 1.461 |
Quantity | 100 g |
UN Number | UN2733 |
Beilstein | 106924 |
Sensitivity | Air sensitive |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
10255056 | Certificate of Analysis | 17/05/2025 | A13352.22,A13352.0B,A13352.18,A13352.30 |
10223154 | Certificate of Analysis | 30/08/2024 | A13352.22,A13352.0B,A13352.18,A13352.30 |
10207569 | Certificate of Analysis | 13/07/2023 | A13352.22,A13352.0B,A13352,A13352.18,A13352.30 |
10207690 | Certificate of Analysis | 13/07/2023 | A13352.22,A13352.0B,A13352,A13352.18,A13352.30 |
J3262A | Certificate of Analysis | 22/11/2021 | A13352.22,A13352.0B,A13352.18,A13352.30 |
Safety and Handling


- 1-Methyl-4-piperidone
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 1-Methyl-4-piperidone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.