Learn More
1-Methyl-4-piperidinone, Sigma-Aldrich™
Product Code. 15861201
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity Unit Size
15861201 100 mL 100mL
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15861201 Supplier Sigma Aldrich Supplier No. 8208280100
 Restricted Item
Forventet forsendelse: 18-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 1445-73-4
Density 0.97 g/cm3 at 20°C
Boiling Point 60°C to 62°C/19 hPa
MDL Number MFCD00006191
Quantity 100 mL
Synonym 1-Methyl-4-piperidinone, N-Methyl-γ-piperidone, 1-Methyl-4-piperidinone
Molecular Weight (g/mol) 113.16
Grade Research