Få mere at vide
1-Methyl-4-nitro-3-propyl-1H-pyrazole-5-carboxamide, Mikromol™
Artikelnummer. 30049871
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049871 25 mg
17983336 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049871 Leverandør Mikromol Leverandørnr. MM0254.270025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Methyl-4-nitro-3-propyl-1H-pyrazole-5-carboxamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
CAS 139756-01-7
Analyt- eller komponentnavne 1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C8 H12 N4 O3
Certificeringer/compliance ISO 9001
Synonym 1H-Pyrazole-5-carboxamide, 1-methyl-4-nitro-3-propyl-, 1-Methyl-4-nitro-3-propyl-1H-pyrazole-5-carboxamide, 1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
InChI formel InChI=1S/C8H12N4O3/c1-3-4-5-6(12(14)15)7(8(9)13)11(2)10-5/h3-4H2,1-2H3,(H2,9,13)
SMIL CCCc1nn(C)c(C(=O)N)c1[N+](=O)[O-]
IUPAC navn 2-methyl-4-nitro-5-propylpyrazole-3-carboxamide
Molekylvægt (g/mol) 212.21
Mængde 25 mg
Formel vægt 212.0909 g/mol