Få mere at vide
1-Methyl-4-nitro-1H-imidazole-5-carboxylic Acid, TRC
Artikelnummer. 30476450
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476450 5 g
30473506 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30476450 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3237485G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54828-05-6
Molekylær formel C5 H5 N3 O4
Molekylvægt (g/mol) 171.11
IUPAC navn 3-methyl-5-nitroimidazole-4-carboxylic acid
SMIL Cn1cnc(c1C(=O)O)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 1-Methyl-4-nitro-1H-imidazole-5-carboxylic Acid
Mængde 5 g
InChI formel InChI=1S/C5H5N3O4/c1-7-2-6-4(8(11)12)3(7)5(9)10/h2H,1H3,(H,9,10)
Formel vægt 171.028