Få mere at vide
1-Methyl-4-hydroxypiperidine-d4, TRC
Artikelnummer. 30465572
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30465572 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465572 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31287250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-4-hydroxypiperidine-d4
CAS 1189499-82-8
Molekylær formel C6H9D4NO
Mængde 50 mg
InChI formel InChI=1S/C6H13NO/c1-7-4-2-6(8)3-5-7/h6,8H,2-5H2,1H3/i4D2,5D2
SMIL [2H]C1([2H])CC(O)CC([2H])([2H])N1C
IUPAC navn 2,2,6,6-tetradeuterio-1-methylpiperidin-4-ol
Molekylvægt (g/mol) 119.2
Formel vægt 119.1248