Learn More
1-Methyl-3-piperazinone, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
It is a useful piperazine and also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful piperazine and also used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is a useful piperazine and also used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Melting Point | 84°C to 90°C |
Quantity | 250 mg |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 114.15 |
Percent Purity | 95% |
Chemical Name or Material | 1-Methyl-3-piperazinone |