missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-piperazinone, 95%, Thermo Scientific Chemicals

Catalog Number 15444078
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
34770-60-0
C5H10N2O
114.15
MFCD11870124
KVIZTDNKHOCNAM-UHFFFAOYSA-N
1-methyl-3-oxopiperazine, 1-methyl-3-piperazinone, 1-methylpiperazin-3-one, piperazinone, 4-methyl, pubchem23318, 1-methyl-3-piperizinone, 4-methyl-2-piperazinone, 4-methyl-piperazin-2-one, 2-piperazinone,4-methyl
13704283
4-methylpiperazin-2-one
CN1CCNC(=O)C1
This item is not returnable. View return policy
34770-60-0
C5H10N2O
114.15
MFCD11870124
KVIZTDNKHOCNAM-UHFFFAOYSA-N
1-methyl-3-oxopiperazine, 1-methyl-3-piperazinone, 1-methylpiperazin-3-one, piperazinone, 4-methyl, pubchem23318, 1-methyl-3-piperizinone, 4-methyl-2-piperazinone, 4-methyl-piperazin-2-one, 2-piperazinone,4-methyl
13704283
4-methylpiperazin-2-one
CN1CCNC(=O)C1

It is a useful piperazine and also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful piperazine and also used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 84°C to 90°C
Quantity 250 mg
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 114.15
Percent Purity 95%
Chemical Name or Material 1-Methyl-3-piperazinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'