missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Methyl-3-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63787.06

Additional Details : CAS Number : 18391-77-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15470669

  • 5957.57 DKK / 5g


Estimated Shipment date
from Supplier 15-01-2019
Add to basket

Specifications

Chemical Identifiers

18391-77-0
206.263
OADQZAOQKGDEEA-UHFFFAOYSA-N
854388
CN1CC(=O)N(C1=S)C2=CC=CC=C2
C10H10N2OS
MFCD00464059
1-methyl-3-phenyl-2-thioxoimidazolidin-4-one, 4-imidazolidinone, 1-methyl-3-phenyl-2-thioxo, 1-methyl-3-phenyl-2-sulfanylidene-imidazolidin-4-one, 1-methyl-3-phenyl-2-thioxo-1,3-diazolidin-4-one, 1-methyl-3-phenyl-2-thioxo-4-imidazolidinone
1-methyl-3-phenyl-2-sulfanylideneimidazolidin-4-one

Specifications

1-Methyl-3-phenyl-2-thiohydantoin
18391-77-0
MFCD00464059
1-methyl-3-phenyl-2-thioxoimidazolidin-4-one, 4-imidazolidinone, 1-methyl-3-phenyl-2-thioxo, 1-methyl-3-phenyl-2-sulfanylidene-imidazolidin-4-one, 1-methyl-3-phenyl-2-thioxo-1,3-diazolidin-4-one, 1-methyl-3-phenyl-2-thioxo-4-imidazolidinone
CN1CC(=O)N(C1=S)C2=CC=CC=C2
206.263
206.26
95%
5g
C10H10N2OS
168827
OADQZAOQKGDEEA-UHFFFAOYSA-N
1-methyl-3-phenyl-2-sulfanylideneimidazolidin-4-one
854388
Solid
Videos
SDS