Få mere at vide
1-Methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]guanidine Dihydrochloride, Mikromol™
Beskrivelse
1-Methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]guanidine Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1-Methyl-3-[2-([(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl)ethyl]guanidine Dihydrochloride |
| CAS | 59660-24-1 |
| Analyt- eller komponentnavne | 1-Methyl-3-[2-([(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl)ethyl]guanidine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H17 N5 S . 2 Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Cimetidine Hydrochloride Imp. D (EP), Cimetidine Imp. D (EP), 1-Methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]guanidine Dihydrochloride, Cimetidine Hydrochloride Imp. D (EP) as Dihydrochloride, SKF-92408A2, Ph Eur Cimetidine Impurity D, Cimetidine Hydrochloride Impurity D, Cimetidine Impurity D as Dihydrochloride, Cimetidine Impurity D |
| InChI formel | InChI=1S/C9H17N5S.2ClH/c1-7-8(14-6-13-7)5-15-4-3-12-9(10)11-2;;/h6H,3-5H2,1-2H3,(H,13,14)(H3,10,11,12);2*1H |
| SMIL | Cl.Cl.CNC(=N)NCCSCc1nc[nH]c1C |
| IUPAC navn | 1-Methyl-3-[2-([(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl)ethyl]guanidine Dihydrochloride |
| Vis mere |