Få mere at vide
1-Methyl-2-pyrrolidinone-d3, TRC
Artikelnummer. 30478759
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30478759 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478759 Leverandør Toronto Research Chemicals Leverandørnr. TRCM326122250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Methyl-2-pyrrolidinone-d3
CAS 933-86-8
Molekylær formel C5H6D3NO
Mængde 250 mg
Synonym 1-Methyl-5-pyrrolidinone-d3, 1-Methylazacyclopentan-2-one-d3, 1-Methylpyrrolidone-d3, AgsolEx 1-d3, M-Pyrol-d3, Microposit 2001-d3, N 0131-d3, N-Methyl-2-ketopyrrolidine-d3, N-Methyl-γ-butyrolactam-d3, N-Methylbutyrolactam-d3, N-Methylpyrrolidone-d3, NMP-d3, NSC 4594-d3, Pharmasolve-d3, Pyrol M-d3, SL 1332-d3
InChI formel InChI=1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3/i1D3
SMIL O=C1CCCN1C([2H])([2H])[2H]
Molekylvægt (g/mol) 102.15