missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals

Catalog Number p-4812625
Click to view available options
Quantity:
5 L
25 L
872-50-4
C5H9NO
99.13
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
CN1CCCC1=O
872-50-4
C5H9NO
99.13
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
CN1CCCC1=O

1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -24.0°C
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 5 L
Flash Point 91°C
Packaging Metal Drum
Refractive Index 1.4690 to 1.4710
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Merck Index 15,619
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 98%
Grade Pure
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Methyl-2-pyrrolidone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'