missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-2-imidazolecarboxaldehyde, 97%, Thermo Scientific Chemicals Product Code.: 10262672

1-Methyl-2-imidazolecarboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. p-2689558 Shop All Thermo Scientific Chemicals Products
1 g, Glass bottle, 1g
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10262672

Brand: Thermo Scientific Acros 369850010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13750-81-7
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01321308
InChI Key UEBFLTZXUXZPJO-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-2-carbaldehyde, 1-methyl-2-imidazolecarboxaldehyde, 1-methylimidazole-2-carboxaldehyde, 1h-imidazole-2-carboxaldehyde, 1-methyl, 2-formyl-1-methylimidazole, imidazole-2-carboxaldehyde, 1-methyl, pubchem7614, zlchem 1231, 2-formyl-l-methylimidazole, acmc-209ce6
PubChem CID 139575
IUPAC Name 1-methylimidazole-2-carbaldehyde
SMILES CN1C=CN=C1C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 35°C to 39°C
Color Colorless to Yellow
Boiling Point 70°C to 74°C (1.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 110.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Methyl-2-imidazolecarboxaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0430665 Certificate TypeCertificate of Analysis Date11/12/2023 Product Code369850010,369850050
Lot NumberA0429652 Certificate TypeCertificate of Analysis Date10/11/2023 Product Code369850010,369850050
Lot NumberA0391946 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code369850010,369850050
Lot NumberA0446456 Certificate TypeCertificate of Analysis Date24/10/2022 Product Code369850010,369850050
Lot NumberA0392799 Certificate TypeCertificate of Analysis Date19/07/2022 Product Code369850010,369850050
compliance-icons
Product Identifier
  • 1-Methyl-2-imidazolecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-2-imidazolecarboxaldehyde, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.